Amplite® Colorimetric NAD/NADH Ratio Assay Kit
Supplier: AAT BIOQUEST
15273
AATB15273EA 537 EUR
AATB15273
Amplite® Colorimetric NAD/NADH Ratio Assay Kit
Assays Enzyme Assays
Nicotinamide adenine dinucleotide (NAD+) and nicotinamide adenine dinucleotide phosphate (NADP+) are two important cofactors found in cells.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.