ReadiView™ Blue Real-Time PCR Visualisation Dye *200X*
Supplier: AAT BIOQUEST
17300
AATB17300EA 136 EUR
AATB17300
ReadiView™ Blue Real-Time PCR Visualisation Dye *200X*
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
ReadiView™ Blue Real-Time PCR Visualisation Dye is a 200X concentrated, ready-to-use, inert dye that enhances the visibility of real-time PCR reactions for more accurate pipetting and plate loading.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.