Helixyte™ Green Fluorometric dsDNA Quantitation kit *High Sensitivity*
Supplier: AAT BIOQUEST
17651
AATB17651EA 789 EUR
AATB17651
Helixyte™ Green Fluorometric dsDNA Quantitation kit *High Sensitivity*
Assays Nucleic Acid Assays
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.