ReadiLink™ xtra Rapid iFluor® 350 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
Supplier: AAT BIOQUEST
1950
AATB1950EA 292 EUR
AATB1950
ReadiLink™ xtra Rapid iFluor® 350 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
Protein Linking and Labeling Systems
ReadiLink™ xtra rapid antibody labelling kits require essentially only two simple mixing steps without a column purification needed.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.