Annexin V, XFD488 Conjugate
Supplier: AAT BIOQUEST
20092
AATB20092EA 307 EUR
AATB20092
Annexin V, XFD488 Conjugate
Assays Cellular Assays
XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
  • High stability
  • In-house quality control
  • Multiple research applications
Caution: For research use only.