Cell Meter™ Live Cell Caspase 9 Binding Assay Kit *Green Fluorescence*
Supplier: AAT BIOQUEST
20117
AATB20117EA 460 EUR
AATB20117
Cell Meter™ Live Cell Caspase 9 Binding Assay Kit *Green Fluorescence*
Assays Cellular Assays
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.