Nile red ≥95%

Supplier: AAT BIOQUEST
Warning

22190
AATB22190EA 120 EUR
AATB22190
Nile red ≥95%
Nile red

Nile red (also known as Nile blue oxazone) is a lipophilic stain.

Formula: C₂₀H₁₈N₂O₂
MDL Number: MFCD00011639
CAS Number: 7385-67-3
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR