Tetracysteine FLASH Diacetate *2 mM DMSO Solution*
Supplier: AAT BIOQUEST
22334
AATB22334EA 568 EUR
AATB22334
Tetracysteine FLASH Diacetate *2 mM DMSO Solution*
Assays Protein Assays
FLASH diacetate is a cell-permeable version of FlASH, which is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
Caution: For research use only.