Cell Meter™ Cell Viability Assay Kit *Red Fluorescence*
Supplier: AAT BIOQUEST
22783
AATB22783EA 460 EUR
AATB22783
Cell Meter™ Cell Viability Assay Kit *Red Fluorescence*
Assays Cellular Assays
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours
Caution: For research use only.