Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Green Fluorescence*
Supplier: AAT BIOQUEST
22900
AATB22900EA 307 EUR
AATB22900
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Green Fluorescence*
Assays Cellular Assays
Reactive oxygen species (ROS) are natural byproducts of the normal metabolism of oxygen and play important roles in cell signaling.
  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Suitable for all major fluorescence platforms
  • Results with only 1 hour incubation
Caution: For research use only.