Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*
Supplier: AAT BIOQUEST
22971
AATB22971EA 460 EUR
AATB22971
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*
Assays Cellular Assays
Mitochondria are major producers of cellular superoxide.
  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimised for microplate readers but suitable for all major fluorescence platforms
  • results with only 1 hour incubation
Caution: For research use only.
Order Now
SPECIFICATIONS
Environmentally Preferable