Amplite® Lactose Quantitation Kit
Supplier: AAT BIOQUEST
40008
AATB40008EA 618 EUR
AATB40008
Amplite® Lactose Quantitation Kit
Assays Cellular Assays
Designed for consistent and dependable results. The Amplite® lactose quantitation kit employs an enzymatic assay to determine lactose concentration, converting lactose into galactose and glucose.
  • Oxidation of galactose yields a colorimetric (570 nm) and fluorometric (Ex/Em = 540/590 nm) product, directly proportional to the initial lactose level
  • Consistent results
  • Six-month shelf life
This versatile kit enables lactose measurement in diverse biological samples, including serum, plasma, other body fluids, food, and growth media.
Delivery information: Shipped with blue ice.
Caution: Research use only.