DBCO-PEG4-NHS Ester for biomolecular labeling

Supplier: AAT BIOQUEST
Warning

4530
AATB4530EA 152 EUR
AATB4530
DBCO-PEG4-NHS Ester for biomolecular labeling
DBCO-PEG4-NHS Ester

Cu-free click reaction, i.e., the strain-promoted alkyne-azide cycloaddition (SPAAC) is a bioorthogonal reaction utilizing a pair of reagents, cyclooctynes and azides that exclusively and efficiently react with each other while remain inert to naturally occurring functional groups such as hydroxy, amino and thiol groups.


  • Improved crosslinking efficiency
  • High water solubility

Formula: C₃₄H₃₉N₃O₁₀
MDL Number: MFCD26793797
CAS Number: 1353016-71-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f7f35b6e87965b85e9c2923cf32789c6f956ef2&vid=8804e6d5727b75e7dcd78d02bd0328679c634daa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR