Rhodamine 6G 90

Supplier: AAT BIOQUEST
Warning

Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

70
AATB70EA 120 EUR
AATB70
Rhodamine 6G 90
Rhodamine 6G

Rhodamine 6G is also called Rhodamine 590, R6G, Basic Rhodamine Yellow, or C.I.

Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
MDL Number: MFCD00012665
CAS Number: 989-38-8
ADR: 5 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR