Digoxigenin (DIG) azide, FastClick™
Supplier: AAT BIOQUEST
72800
AATB72800EA 266 EUR
AATB72800
Digoxigenin (DIG) azide, FastClick™
Digoxigenin (DIG) azide
FastClick™ Digoxigenin (DIG) Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and DIG hapten (as the detection tag) for developing DIG-based probes.
  • Stable for 24 months
Quality tested in-house
Caution: For Research Use Only