Anti-CD68 Mouse Monoclonal Antibody [clone: 404CT9.2.5]

Supplier: Abgent
AM1996A
ABGEAM1996AEA 354 EUR
ABGEAM1996A
Anti-CD68 Mouse Monoclonal Antibody [clone: 404CT9.2.5]
Antibodies
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 404CT9.2.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR