Anti-K1199 Rabbit Polyclonal Antibody

Supplier: Abgent
AP10964B
ABGEAP10964BEA 354 EUR
ABGEAP10964B
Anti-K1199 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: K1199
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR