Anti-CE164 Rabbit Polyclonal Antibody

Supplier: Abgent
AP10981A
ABGEAP10981AEA 354 EUR
ABGEAP10981A
Anti-CE164 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CE164
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CEP164
Environmentally Preferable
Antigen name Centrosomal protein 164kDa
Antigen synonyms MEK1|MAPKK1|CFC3|PRKMK1|MKK1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope N-Terminal (1–100 aa)
Cross adsorption No
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR