Anti-SPACA5 Rabbit Polyclonal Antibody

Supplier: Abgent
AP11801C
ABGEAP11801CEA 354 EUR
ABGEAP11801C
Anti-SPACA5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPACA5 (sperm acrosome associated 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR