Anti-PTGR2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP12869A
ABGEAP12869AEA 354 EUR
ABGEAP12869A
Anti-PTGR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTGR2 (prostaglandin reductase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR