Anti-BCL2L14 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1304A
ABGEAP1304AEA 354 EUR
ABGEAP1304A
Anti-BCL2L14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BCL2L14 (BCL2-like 14 (apoptosis facilitator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: BH3 domain
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR