Anti-FGD2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP13191B
ABGEAP13191BEA 354 EUR
ABGEAP13191B
Anti-FGD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGD2 (FYVE, RhoGEF and PH domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR