Anti-HM13 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14392B
ABGEAP14392BEA 354 EUR
ABGEAP14392B
Anti-HM13 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HM13 (histocompatibility (minor) 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR