Anti-DNMT3A Rabbit Polyclonal Antibody

Supplier: Abgent
AP16264B
ABGEAP16264BEA 354 EUR
ABGEAP16264B
Anti-DNMT3A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR