Anti-FBXO46 Rabbit Polyclonal Antibody

Supplier: Abgent
AP16343B
ABGEAP16343BEA 354 EUR
ABGEAP16343B
Anti-FBXO46 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FBXO46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR