Anti-RNASE11 Rabbit Polyclonal Antibody

Supplier: Abgent
AP17847C
ABGEAP17847CEA 354 EUR
ABGEAP17847C
Anti-RNASE11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNASE11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol RNASE11
Environmentally Preferable
Antigen name Ribonuclease, RNase A family, 11 (non-active)
Antigen synonyms HEL-S-84p|RAJ1|C14orf6
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 122651
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flow cytometry Yes
Epitope internal
Immunogen This RNASE11 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 68-95 amino acids from the Central region of human RNASE11.
Amino acid number 68 to 95
Sequence YNDPKGNSSGNDKC
Molecular weight 22.427 kDa
Purification Protein A & Peptide affinity purification
Concentration 0.47
Storage buffer pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR