Anti-LCP1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20707C
ABGEAP20707CEA 354 EUR
ABGEAP20707C
Anti-LCP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR