Anti-CDKN1B Rabbit Polyclonal Antibody

Supplier: Abgent
AP3192A
ABGEAP3192AEA 354 EUR
ABGEAP3192A
Anti-CDKN1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR