Anti-ACE2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6020D
ABGEAP6020DEA 354 EUR
ABGEAP6020D
Anti-ACE2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR