Anti-SPHK1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7237A
ABGEAP7237AEA 354 EUR
ABGEAP7237A
Anti-SPHK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR