Anti-GABRA4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9298C
ABGEAP9298CEA 354 EUR
ABGEAP9298C
Anti-GABRA4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GABRA4 (gamma-aminobutyric acid (GABA) A receptor, alpha 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GABRA4
Environmentally Preferable
Antigen name Gamma-aminobutyric acid (GABA) A Receptor alpha 4
Antigen synonyms Gamma-aminobutyric acid receptor subunit alpha-4|GABRA4|Gamma aminobutyric acid GABA A receptor alpha 4|Gamma aminobutyric acid receptor alpha 4 subunit|Gamma aminobutyric acid receptor subunit alpha 4|GABAA receptor subunit alpha 4|GABRA 4|GBRA4_HUMAN.|GABAA receptor subunit alpha-4|Gamma aminobutyric acid A receptor alpha 4|GABR A4
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 134187
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flow cytometry Yes
Epitope Center
Immunogen This POU5F2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 203-231 amino acids from the Central region of human POU5F2.
Sequence ERRIGNSLEKFFQRC
Purification Protein A & Peptide affinity purification
Concentration 0,5 mg/ml
Storage buffer PBS with 0,09% (W/V) sodium azide, pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR