Anti-atg4b Rabbit Polyclonal Antibody

Supplier: Abgent
AZB10037A
ABGEAZB10037AEA 354 EUR
ABGEAZB10037A
Anti-atg4b Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: atg4b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR