Anti-BHMT Mouse Polyclonal Antibody

Supplier: Abnova
H00000635-B01P
ABNOH00000635-B01PEA 397 EUR
ABNOH00000635-B01P
Anti-BHMT Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BHMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR