Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 4A9]

Supplier: Abnova
H00000944-M08
ABNOH00000944-M08EA 397 EUR
ABNOH00000944-M08
Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 4A9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFSF8.

Type: Primary
Antigen: TNFSF8
Clonality: Monoclonal
Clone: 4A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR