Anti-SOCS3 Mouse Monoclonal Antibody [clone: 1E4]

Supplier: Abnova
H00009021-M02
ABNOH00009021-M02EA 397 EUR
ABNOH00009021-M02
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 1E4]
Antibodies
Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR