Anti-PDIA4 Mouse Monoclonal Antibody [clone: 2H3]

Supplier: Abnova
H00009601-M02
ABNOH00009601-M02EA 397 EUR
ABNOH00009601-M02
Anti-PDIA4 Mouse Monoclonal Antibody [clone: 2H3]
Antibodies
Type: Primary
Antigen: PDIA4
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR