Anti-NCAM1 Mouse Monoclonal Antibody (Biotin) [clone: B-A19]

Supplier: Abnova
MAB13716
ABNOMAB13716EA 402 EUR
ABNOMAB13716
Anti-NCAM1 Mouse Monoclonal Antibody (Biotin) [clone: B-A19]
Antibodies
Mouse monoclonal antibody raised against human NCAM1.

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: B-A19
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol NCAM1
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone B-A19
Reactivity Human
Host Mouse
Gene ID 4684
Isotype IgG1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR