Anti-COX1 Mouse Monoclonal Antibody

Supplier: Abnova
MAB3797
ABNOMAB3797EA 523 EUR
ABNOMAB3797
Anti-COX1 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against PTGS1.

Type: Primary
Antigen: COX1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Guinea Pig, Human, Rabbit, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR