Anti-TBL1Y Mouse Monoclonal Antibody

Supplier: Abnova
MAB8712
ABNOMAB8712EA 402 EUR
ABNOMAB8712
Anti-TBL1Y Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against full length recombinant TBL1Y.

Recommended Dilutions: Western Blot: 1:100-1:2000; ELISA: 1:5000-1:20000

Type: Primary
Antigen: TBL1Y
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR