Anti-HCG_1776007 Chicken Polyclonal Antibody

Supplier: Abnova
PAB29085
ABNOPAB29085EA 779 EUR
ABNOPAB29085
Anti-HCG_1776007 Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody raised against recombinant Human hCG_1776007.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: HCG_1776007
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol HCG_1776007
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Chicken
Gene ID 146713|52897
Isotype IgY
ImmunoChemistry Yes
Immunogen A KLH-conjugated synthetic peptide corresponding to the hCG_1776007 gene product, shared between the human (NP_001076044, NCBI) and mouse (NP_001020102, NCBI) sequences.
Format Liquid
Purification Antigen affinity purification
Cross adsorption No
Storage buffer PBS, pH 7,2 (50% glycerol, 0,02% sodium azide)
Storage temperature Store at 4 °C and avoid from light. Do not freeze unless for long term storage, store at –20 °C with 50% glycerol. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR