Anti-GALNT6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB30093
ABNOPAB30093EA 523 EUR
ABNOPAB30093
Anti-GALNT6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of human GALNT6.

Type: Primary
Antigen: GALNT6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GALNT6
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Reactivity Human
Host Rabbit
Gene ID 11226
Isotype
Western blot Yes
ImmunoChemistry Yes
Sequence MNNLRDSMPKLQIRAPEAQQTLFSINQSCLPGFYTPAELKPFWERPPQDP

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR