Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
ACE-111-0546EA 977 EUR
ACE-111-0546 ACE-111-1546 ACE-111-0246 ACE-111-0302 ACE-111-0303 ACE-111-0346 ACE-111-0501 ACE-111-0502 ACE-111-0503 ACE-111-0504 ACE-111-1001 ACE-111-1002 ACE-111-1003 ACE-111-1004 ACE-111-1046 ACE-111-1202 ACE-111-1203 ACE-111-1204 ACE-111-1246 ACE-111-1501 ACE-111-1502 ACE-111-1503 ACE-111-1504 ACE-111-3502 ACE-111-3546 ACE-111-7502 ACE-111-7503 ACE-111-7504 ACE-111-7546 ACE-111-0103GD ACE-111-0101GD ACE-111-0102GD ACE-111-10003 ACE-111-10005 ACE-111-15003 ACE-111-05003 ACE-111-05005S ACE-111-005003GD ACE-111-1500075 ACE-111-3346 ACE1111046MVK ACE1111546MVK ACETACE-111-15003S ACE-111-1539
Avantor® ACE® C18, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information