Avantor® ACE® C18-PFP, HPLC Columns, 5 µm
Supplier: Avantor
ACE-1210-0102GDEA 323 EUR
ACE-1210-0102GD ACE-1210-0103GD ACE-1210-0110GD ACE-1210-0302 ACE-1210-0346 ACE-1210-0502 ACE-1210-0503 ACE-1210-0510 ACE-1210-0520 ACE-1210-0530 ACE-1210-0546 ACE-1210-1002 ACE-1210-1003 ACE-1210-1010 ACE-1210-1020 ACE-1210-1030 ACE-1210-1046 ACE-1210-1204 ACE-1210-1220 ACE-1210-1246 ACE-1210-1502 ACE-1210-1503 ACE-1210-1504 ACE-1210-1510 ACE-1210-1520 ACE-1210-1530 ACE-1210-1546 ACE-1210-2502 ACE-1210-2503 ACE-1210-2504 ACE-1210-2508 ACE-1210-2510 ACE-1210-2520 ACE-1210-2530 ACE-1210-2546 ACE-1210-7520 ACE-1210-7530 ACE-1210-7546 ACE-1210-3020 ACE-1210-3030 ACE12100120GD ACE12102546MVK
Avantor® ACE® C18-PFP, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C18-PFP phase utilises a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 µm to 10 µm) and complementary phases
  • Also useful for analytes with π-bonding or
More Product Information