2-Acetylpyridine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(Pyridin-2-yl)ethanone, Methyl 2-pyridyl ketone

102680250 102681000 102685000
ACRO102680250EA 57.7 EUR
ACRO102680250 ACRO102681000 ACRO102685000
2-Acetylpyridine 98%
2-Acetylpyridine
Formula: C₇H₇NO
Boiling Pt: 188…189 °C (760 mmHg)
Melting Pt: 8…10 °C
Density: 1,08 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00006303
CAS Number: 1122-62-9
EINECS: 214-355-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5250 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR