2-(Aminomethyl)pyridine 99%

Supplier: Thermo Fisher Scientific
Danger

104070250 104071000 104075000
ACRO104070250EA 87.5 EUR
ACRO104070250 ACRO104071000 ACRO104075000
2-(Aminomethyl)pyridine 99%
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
Boiling Pt: 82…85 °C (12 mmHg)
Melting Pt: –20 °C
Density: 1,04 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00006360
CAS Number: 3731-51-9
EINECS: 223-090-5
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR