Benzylacetone 99%

Supplier: Thermo Fisher Scientific
Warning

105830050 105832500 105830010
ACRO105830050EA 26.2 EUR
ACRO105830050 ACRO105832500 ACRO105830010
Benzylacetone 99%
Benzylacetone
Formula: (C₆H₅)CH₂CH₂C(O)CH₃
Boiling Pt: 235…237 °C (760 mmHg)
Melting Pt: -13 °C
Density: 0,9875 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00008790
CAS Number: 2550-26-7
EINECS: 219-847-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.985 to 0.99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR