Chlorocyclohexane 98%

Supplier: Thermo Fisher Scientific
Warning

111300010 111301000 111302500
ACRO111300010EA 104 EUR
ACRO111300010 ACRO111301000 ACRO111302500
Chlorocyclohexane 98%
Chlorocyclohexane
Formula: C₆H₁₁Cl
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00003822
CAS Number: 542-18-7
EINECS: 208-806-6
UN: 1993
ADR: 3,III
Merck Index: 13,02761

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
Water ≤0.05 %
GC ≥97.5 %
Color scale ≤10 APHA
Water soluble titr. acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR