1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: CMC

111360050 111360250
ACRO111360050EA 139 EUR
ACRO111360050 ACRO111360250
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate 95%
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate

Appearance: White to light yellow Powder

Formula: C₂₁H₃₃N₃O₄S
Melting Pt: 110…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00011979
CAS Number: 2491-17-0

Order Now

Specification Test Results

Appearance White to light yellow Powder and/or chunks
Infrared spectrum Conforms
Melting point 112 to 120 °C
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR