Dehydroacetic acid 98%

Supplier: Thermo Fisher Scientific
Warning

111940050 111941000 111945000
ACRO111940050EA 26.2 EUR
ACRO111940050 ACRO111941000 ACRO111945000
Dehydroacetic acid 98%
Dehydracetic acid
Formula: C₈H₈O₄
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 109…112 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885

Order Now

Specification Test Results

Appearance Cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 109 to 112 °C
Sulfated ash ≤0.1 % (700 °C, 2 h)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR