1,4-Diaminobutane 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Putrescine, 1,4-Butanediamine, Tetramethylendiamine

112120250 112121000 112125000 112120025
ACRO112120250EA 56.9 EUR
ACRO112120250 ACRO112121000 ACRO112125000 ACRO112120025
1,4-Diaminobutane 99%
1,4-Diaminobutane
Formula: C₄H₁₂N₂
Boiling Pt: 158…160 °C (760 mmHg)
Melting Pt: 27 °C
Density: 0,877 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008235
CAS Number: 110-60-1
EINECS: 203-782-3
UN: 2923
ADR: 8,II
Merck Index: 12,08134

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4590 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +21° to +25° (22°C, 589 nm) (c=1.2, 1 N HCl)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR